Product Details
- SNP ID
-
rs13481477
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:58267527 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTGTGGTGCAAAGAGGTAGCCTC[A/G]GCATCACATTCCACACCTGGTGAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Clec14a
PubMed Links
Gene Details
- Gene
- Clec14a
- Gene Name
- C-type lectin domain family 14, member a
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025809.5 |
1732 |
Silent Mutation |
|
|
NP_080085.3 |
View Full Product Details