Product Details
- SNP ID
-
rs4172058
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:34397165 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACTCCATCCTTCCCCTGCACTCT[C/T]CACCTAGCCACATAGTACCCATGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kalrn
PubMed Links
Gene Details
- Gene
- Kalrn
- Gene Name
- kalirin, RhoGEF kinase
View Full Product Details