Product Details
- SNP ID
-
rs4167975
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30308841 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCGCGAGCACGCGCAGCTCCCG[C/T]AGGCCCTGGAACACGCCGCGCGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Atp13a3
PubMed Links
Gene Details
- Gene
- Atp13a3
- Gene Name
- ATPase type 13A3
There are no transcripts associated with this gene.
- Gene
- Gp5
- Gene Name
- glycoprotein 5 (platelet)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008148.4 |
1043 |
Silent Mutation |
|
|
NP_032174.2 |
View Full Product Details