Product Details
- SNP ID
-
rs13481082
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:68278917 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAAGCATGCCCAGTGTAAGAGTG[A/G]CCCTCCCATTCCAAGGCCAGCTTGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Ntn1
PubMed Links
Gene Details
- Gene
- Ntn1
- Gene Name
- netrin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008744.2 |
|
Intron |
|
|
NP_032770.2 |
View Full Product Details