Product Details
- SNP ID
-
rs13482234
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:67809100 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAATTCCGTCTTGCTAGGGTCCTC[G/T]GCATTGGCCACATAAAAGTTCAGTA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dock5
PubMed Links
Gene Details
- Gene
- Dock5
- Gene Name
- dedicator of cytokinesis 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177780.3 |
2470 |
Silent Mutation |
|
|
NP_808448.2 |
View Full Product Details