Product Details
- SNP ID
-
rs3669009
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:10607084 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTCCTATTTCACATTCCATAAC[A/G]CTGTAGTATAACATTCAATAATTCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fhit
PubMed Links
Gene Details
- Gene
- Fhit
- Gene Name
- fragile histidine triad gene
There are no transcripts associated with this gene.
View Full Product Details