Product Details
- SNP ID
-
rs13481100
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74465211 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGAGCTGAATAGTGAGTCTGGA[A/G]TGGGTCTACCCAGCCGACACAGTAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rap1gap2
PubMed Links
Gene Details
- Gene
- Rap1gap2
- Gene Name
- RAP1 GTPase activating protein 2
View Full Product Details