Product Details

SNP ID
rs13481819
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:55740984 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCGCAGCAGCAGGACAGAACC[A/G]TGTCTGCAGTCCAGTGGGTTGGGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Pcbd2 PubMed Links

Gene Details

Gene
Pcbd2
Gene Name
pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028281.1 Intron NP_082557.1

View Full Product Details