Product Details

SNP ID
rs13480469
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:4358515 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTAGAATGTGAAGTGAAGTCAC[G/A]GAACAAGATGCTGCTGTTGGGACCT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Akap12 PubMed Links

Gene Details

Gene
Akap12
Gene Name
A kinase (PRKA) anchor protein (gravin) 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031185.3 5291 UTR 3 NP_112462.1
Gene
Zbtb2
Gene Name
zinc finger and BTB domain containing 2
There are no transcripts associated with this gene.

View Full Product Details