Product Details
- SNP ID
-
rs4184376
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:49089467 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTAGCCCTAGGTAGCATTCACAT[C/T]ACCTGAAGAGTCTATTGATTTCTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Myh15
PubMed Links
Gene Details
- Gene
- Myh15
- Gene Name
- myosin, heavy chain 15
View Full Product Details