Product Details
- SNP ID
-
rs13459051
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:130850953 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCAACTTGAGAATCACACTGATC[C/T]GCTCGCAGCCCACACTCACCCATCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pigr
PubMed Links
Gene Details
- Gene
- Pigr
- Gene Name
- polymeric immunoglobulin receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011082.3 |
2553 |
UTR 3 |
|
|
NP_035212.2 |
View Full Product Details