Product Details
- SNP ID
-
rs13480672
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:83014931 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAACTCTGGCCAGGGTGACAGAAT[A/T]TGGCTGATTCCCCCAAGTATCTGAG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Chst11
PubMed Links
Gene Details
- Gene
- Chst11
- Gene Name
- carbohydrate sulfotransferase 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021439.2 |
|
Intron |
|
|
NP_067414.2 |
View Full Product Details