Product Details

SNP ID
rs13476898
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:167134950 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTAGACAGCCTTGTGATCTGT[A/C]AGGCCTGGGCAGTGCTGTCTTTCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Kcnb1 PubMed Links

Gene Details

Gene
Kcnb1
Gene Name
potassium voltage gated channel, Shab-related subfamily, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008420.4 Intron NP_032446.2

View Full Product Details