Product Details

SNP ID
rs13476903
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:168268880 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGTGAGGATGGTGCCAAAAGCTCC[C/G]GGGTTGCGGTCGAAGAAGAACTCGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Kcng1 PubMed Links

Gene Details

Gene
Kcng1
Gene Name
potassium voltage-gated channel, subfamily G, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081134.1 452 Silent Mutation NP_001074603.1

View Full Product Details