Product Details
- SNP ID
-
rs13482531
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:39853416 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAATAGAAAAGGGGATTTCAAGG[A/T]TTCTTCTACAAGGTGAGAAGGCTCT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dpys
PubMed Links
Gene Details
- Gene
- Dpys
- Gene Name
- dihydropyrimidinase
View Full Product Details