Product Details

SNP ID
rs13480802
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:123013422 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTCCTGTAAACTTGTCAGGAC[C/T]GCTTCCGTGTAAGACGGAAGGATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Mon2 PubMed Links

Gene Details

Gene
Mon2
Gene Name
MON2 homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163024.1 4515 Silent Mutation NP_001156496.1
NM_001163025.1 4515 Silent Mutation NP_001156497.1
NM_153395.2 4515 Silent Mutation NP_700444.2

View Full Product Details