Product Details

SNP ID
rs13478738
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:46837420 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTATGTTTACAAGTTGGGTGC[C/T]AAGGAGAACCTGGCAGTGTTACAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cntnap2 PubMed Links

Gene Details

Gene
Cntnap2
Gene Name
contactin associated protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004357.2 Intron NP_001004357.2
NM_025771.3 Intron NP_080047.1

View Full Product Details