Product Details

SNP ID
rs3023345
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:69584488 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCTCAAAGAGTTTAAGAACAG[C/T]GGGTATTATTCCTAAGTCTCCTAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Sos2 PubMed Links

Gene Details

Gene
Sos2
Gene Name
son of sevenless homolog 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135559.1 4885 UTR 3 NP_001129031.1
Gene
Vcpkmt
Gene Name
valosin containing protein lysine (K) methyltransferase
There are no transcripts associated with this gene.

View Full Product Details