Product Details
- SNP ID
-
rs13480967
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:36197132 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTACCATAATATGAAAACCACAA[A/G]TCAAACTATCGTCTAAAATGGCAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tenm2
PubMed Links
Gene Details
- Gene
- Tenm2
- Gene Name
- teneurin transmembrane protein 2
There are no transcripts associated with this gene.
View Full Product Details