Product Details
- SNP ID
-
rs13459052
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:131981704 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACACAGGTAGTGTTTGACAAGAA[C/T]GACTTGGCCAAATACTCAGTGTAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Slc45a3
PubMed Links
Gene Details
- Gene
- Slc45a3
- Gene Name
- solute carrier family 45, member 3
View Full Product Details