Product Details
- SNP ID
-
rs13481997
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:106993050 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGACCTGCACTGATACAGATGGT[A/G]GAAAAAACTTTTTAAAAATGAACCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kif2a
PubMed Links
Gene Details
- Gene
- Kif2a
- Gene Name
- kinesin family member 2A
View Full Product Details