Product Details

SNP ID
rs3680731
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:14354063 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGAATTTATCACTATTAACATTAAA[T/G]TGTTGGTTATATCAAAGAAACATTG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Hecw1 PubMed Links

Gene Details

Gene
Hecw1
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081348.3 Intron NP_001074817.3

View Full Product Details