Product Details
- SNP ID
-
rs6174159
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:106631065 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTCTTTCAGTAGCTTTCTCACC[G/T]AATGCACTCACTTTATCTATCAAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
Bcl2
PubMed Links
Gene Details
- Gene
- Bcl2
- Gene Name
- B cell leukemia/lymphoma 2
View Full Product Details