Product Details
- SNP ID
-
rs4205524
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75296538 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATTCCAACTGCAGCCAAATGTA[A/G]TAAGGTAATTAGGAGTGCAATTCAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Robo2
PubMed Links
Gene Details
- Gene
- Robo2
- Gene Name
- roundabout homolog 2 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175549.4 |
|
Intron |
|
|
NP_780758.3 |
View Full Product Details