Product Details
- SNP ID
-
rs13482990
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:42365242 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTATCTCTTACTTGTATAGATGA[C/T]GTGAGAGGGGTTACAAACAAAACAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ptchd4
PubMed Links
Gene Details
- Gene
- Ptchd4
- Gene Name
- patched domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028474.1 |
|
Intron |
|
|
NP_082750.1 |
View Full Product Details