Product Details
- SNP ID
-
rs13475846
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:45448332 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGAATGAAAGATGTATGCCAAG[C/T]GAGGAAGGCTACAAAACAGCACCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Col5a2
PubMed Links
Gene Details
- Gene
- Col5a2
- Gene Name
- collagen, type V, alpha 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007737.2 |
|
Intron |
|
|
NP_031763.2 |
View Full Product Details