Product Details
- SNP ID
-
rs3686631
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:118051961 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGTTACATGCTACTTTACACACA[A/T]TTGATTTTTTTCACCCTGGGCACTC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dnah11
PubMed Links
Gene Details
- Gene
- Dnah11
- Gene Name
- dynein, axonemal, heavy chain 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010060.3 |
|
Intron |
|
|
NP_034190.3 |
View Full Product Details