Product Details

SNP ID
rs3089240
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:57274483 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAATCTCCTGAACACTGGAAGGAAG[C/G]CTCTTTGGAGCCTGTTCCTCACAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Gria1 PubMed Links

Gene Details

Gene
Gria1
Gene Name
glutamate receptor, ionotropic, AMPA1 (alpha 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113325.2 Intron NP_001106796.1
NM_001252403.1 Intron NP_001239332.1
NM_008165.4 Intron NP_032191.2

View Full Product Details