Gene Transcripts

Gene Symbol
Pcdha13
Entrez Gene ID
116742
Gene Name
protocadherin alpha 13
Gene Aliases
Cnr5
Chromosome Location
Chr. 18: 30036887 - 30215897 on Build Rnor_6.0
UniGene ID
Rn.22308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053934.2 NP_446386.2 1 753
GenBank mRNA AB045586.1 - 1 31
AB113396.1 - 1 753
AY573975.1 - 1 753

View Full Product Details