Gene Transcripts

Gene Symbol
Nup85
Entrez Gene ID
287830
Gene Name
nucleoporin 85
Gene Aliases
Pcnt1
Chromosome Location
Chr. 10: 104118999 - 104137258 on Build Rnor_6.0
UniGene ID
Rn.198900
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025401.1 NP_001020572.1 9 770
GenBank mRNA BC098031.1 - 9 770

View Full Product Details