Gene Transcripts

Gene Symbol
Fam13c
Entrez Gene ID
294565
Gene Name
family with sequence similarity 13, member C
Gene Aliases
Fam13c1, RGD1310149
Chromosome Location
Chr. 20: 19358869 - 19405730 on Build Rnor_6.0
UniGene ID
Rn.1325
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772939.1 XP_008771161.1 9 7953
XM_008772940.1 XP_008771162.1 9 7950
GenBank mRNA FQ212141.1 - 6 700
FQ212194.1 - 1714

View Full Product Details