Gene Transcripts

Gene Symbol
Ikbip
Entrez Gene ID
314730
Gene Name
IKBKB interacting protein
Gene Aliases
IKIP, RGD1305457
Chromosome Location
Chr. 7: 31784405 - 31803195 on Build Rnor_6.0
UniGene ID
Rn.13471
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009430.2 NP_001009430.2 1, 2 175
XM_006241250.2 XP_006241312.1 1, 2 344
GenBank mRNA BC088210.1 - 1, 2 248

View Full Product Details