Gene Transcripts

Gene Symbol
Mageb4
Entrez Gene ID
317271
Gene Name
melanoma antigen family B, 4
Gene Aliases
RGD1561997
Chromosome Location
Chr. X: 61103031 - 61151601 on Build Rnor_6.0
UniGene ID
Rn.190323
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177683.1 NP_001171154.1 3 424
GenBank mRNA BC158636.1 - 3 424

View Full Product Details