Gene Transcripts

Gene Symbol
Txn1
Entrez Gene ID
116484
Gene Name
thioredoxin 1
Gene Aliases
Txn
Chromosome Location
Chr. 5: 75049735 - 75057731 on Build Rnor_6.0
UniGene ID
Rn.29777
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053800.3 NP_446252.1 1 70
GenBank mRNA AF311055.1 - 1 70
BC058454.1 - 155 6886988
FQ209943.1 - 152
FQ210839.1 - 155
FQ216849.1 - 181
FQ217822.1 - 150
FQ221646.1 - 123
X14878.1 - 130

View Full Product Details