Gene Transcripts

Gene Symbol
Retnlg
Entrez Gene ID
288135
Gene Name
resistin-like gamma
Gene Aliases
retn1g
Chromosome Location
Chr. 11: 54512950 - 54514700 on Build Rnor_6.0
UniGene ID
Rn.90340
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181625.1 NP_853656.1 3 290
XM_006248282.2 XP_006248344.1 4 383
GenBank mRNA AJ514932.1 - 3 290

View Full Product Details