Gene Transcripts

Gene Symbol
Ldlrad2
Entrez Gene ID
502992
Gene Name
low density lipoprotein receptor class A domain containing 2
Gene Aliases
RGD1564888
Chromosome Location
Chr. 5: 155914496 - 155922269 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_578500.5 XP_578500.2 2 591

View Full Product Details