Gene Transcripts

Gene Symbol
Mir99ahg
Entrez Gene ID
77994
Gene Name
Mir99a and Mirlet7c-1 host gene (non-protein coding)
Gene Aliases
2810055G20Rik, D730043B02Rik
Chromosome Location
Chr. 16: 77329328 - 77558428 on Build GRCm38
UniGene ID
Mm.394688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015543.2 4 563
GenBank mRNA AK012955.1 - 436
AK021336.1 - 607
AK033222.1 - 168
AK048031.1 - 618
AK083771.1 - 4 564
AK144299.1 - 583
AK148800.1 - 504


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB514463.2 - 2 100 Chr. 16 - 77500585 - 77594924 [+]
GenBankmRNA BB624333.1 - 2 165 Chr. 16 - 77500508 - 77594975 [+]
GenBankmRNA BB656236.1 - 4 559 Chr. 16 - 77329332 - 77501605 [+]
GenBankmRNA CF553191.1 - 4 601 Chr. 16 - 77329211 - 77594731 [+]
GenBankmRNA BB603112.2 - 2 88 Chr. 16 - 77500585 - 77594958 [+]
GenBankmRNA DV067527.1 - 2 472 Chr. 16 - 77500215 - 77594785 [+]
GenBankmRNA BB647956.1 - 6 618 Chr. 16 - 77236317 - 77501580 [+]
GenBankmRNA BB663466.1 - 5 487 Chr. 16 - 77236319 - 77596954 [+]
GenBankmRNA BE654839.1 - 2 289 Chr. 16 - 77500384 - 77691323 [+]

View Full Product Details