Gene Transcripts
- Gene Symbol
- Tbrg3
- Entrez Gene ID
- 21378
- Gene Name
- transforming growth factor beta regulated gene 3
- Gene Aliases
- TB-11
- Chromosome Location
- Chr. 15: 82890483 - 82898461 on Build GRCm38
- UniGene ID
- Mm.417272
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027799.1 |
|
1 |
564 |
|
GenBank mRNA |
AK090121.1 |
- |
|
1889 |
|
BC095996.1 |
- |
1 |
471 |
|
View Full Product Details