Gene Transcripts

Gene Symbol
Rian
Entrez Gene ID
75745
Gene Name
RNA imprinted and accumulated in nucleus
Gene Aliases
5530401N18Rik, C130089L09Rik, Meg8
Chromosome Location
Chr. 12: 109603945 - 109661716 on Build GRCm38
UniGene ID
Mm.293263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028261.1 16 2294
GenBank mRNA AF498294.1 - 6 1071
AF498295.1 - 1 84
AF498296.1 - 1 84
AK017440.1 - 6 1023
EU434919.1 - 21 1933


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ830597.1 - 3 302 Chr. 12 - 109651011 - 109656836 [+]
GenBankmRNA AW210163.1 - 3 413 Chr. 12 - 109650888 - 109655933 [+]
GenBankmRNA CF551655.1 - 6 534 Chr. 12 - 109646681 - 109659792 [+]
GenBankmRNA CB574713.1 - 3 649 Chr. 12 - 109650679 - 109658120 [+]
GenBankmRNA BQ833696.1 - 3 497 Chr. 12 - 109650804 - 109655918 [+]
GenBankmRNA CB574817.1 - 6 552 Chr. 12 - 109646670 - 109658125 [+]
GenBankmRNA BI734352.1 - 7 567 Chr. 12 - 109646666 - 109655931 [+]
GenBankmRNA BU058990.1 - 3 611 Chr. 12 - 109650690 - 109656822 [+]
GenBankmRNA BX529056.1 - 3 414 Chr. 12 - 109650887 - 109655911 [+]
GenBankmRNA CB192724.1 - 5 394 Chr. 12 - 109649221 - 109660888 [+]
GenBankmRNA BQ827918.1 - 3 292 Chr. 12 - 109651013 - 109659753 [+]

View Full Product Details