Gene Transcripts
- Gene Symbol
- SNORA72
- Entrez Gene ID
- 26775
- Gene Name
- small nucleolar RNA, H/ACA box 72
- Gene Aliases
- RNU72, U72
- Chromosome Location
- Chr. 8: 98042086 - 98042217 on Build GRCh38
- UniGene ID
- Hs.684855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002581.1 |
|
1 |
16 |
|
GenBank mRNA |
Y11167.1 |
- |
1 |
16 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK128768.1 |
- |
2 |
1515 |
|
Chr. 8 - 98041726 - 98044121 [-] |
View Full Product Details