Gene Transcripts
- Gene Symbol
- TTTY6
- Entrez Gene ID
- 84672
- Gene Name
- testis-specific transcript, Y-linked 6 (non-protein coding)
- Gene Aliases
- LINC00127, NCRNA00127, TTTY6A, TTY6
- Chromosome Location
- Chr. Y: 22439590 - 22441459 on Build GRCh38
- UniGene ID
- Hs.535085
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001527.2 |
|
2 |
185 |
|
- Gene Symbol
- TTTY6B
- Entrez Gene ID
- 441543
- Gene Name
- testis-specific transcript, Y-linked 6B (non-protein coding)
- Gene Aliases
- LINC00128, NCRNA00128, TTTY6, TTY6
- Chromosome Location
- Chr. Y: 22144987 - 22146834 on Build GRCh38
- UniGene ID
- Hs.535085
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002175.3 |
|
2 |
163 |
|
View Full Product Details