Gene Transcripts
- Gene Symbol
- GEMIN8P4
- Entrez Gene ID
- 492303
- Gene Name
- gem nuclear organelle associated protein 8 pseudogene 4
- Gene Aliases
- Chromosome Location
- Chr. 1: 89993265 - 89994966 on Build GRCh38
- UniGene ID
- Hs.550796
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002830.1 |
|
1 |
1323 |
|
GenBank mRNA |
AY726571.1 |
- |
1 |
1314 |
|
View Full Product Details