Gene Transcripts
- Gene Symbol
- PDIA3P1
- Entrez Gene ID
- 171423
- Gene Name
- protein disulfide isomerase family A member 3 pseudogene 1
- Gene Aliases
- ERp60, GRP58P, PDIA3P
- Chromosome Location
- Chr. 1: 147177851 - 147179949 on Build GRCh38
- UniGene ID
- Hs.658566
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002305.1 |
|
1 |
91 |
|
GenBank mRNA |
BM555251.1 |
- |
1 |
35 |
|
View Full Product Details