Gene Transcripts

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
Chromosome Location
Chr. 7: 69598475 - 70793068 on Build GRCh38
UniGene ID
Hs.21631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK055586.1 - 2 336


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA581448.1 - 2 336 Chr. 7 - 70316916 - 70436177 [+]

View Full Product Details