Gene Transcripts
- Gene Symbol
- BCL10
- Entrez Gene ID
- 8915
- Gene Name
- B-cell CLL/lymphoma 10
- Gene Aliases
- CARMEN, CIPER, CLAP, IMD37, c-E10, mE10
- Chromosome Location
- Chr. 1: 85265776 - 85276904 on Build GRCh38
- UniGene ID
- Hs.193516
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098790.1 |
- |
1 |
1336 |
|
View Full Product Details