Gene Transcripts
- Gene Symbol
- SNAP25-AS1
- Entrez Gene ID
- 100131208
- Gene Name
- SNAP25 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 10023812 - 10219506 on Build GRCh38
- UniGene ID
- Hs.658178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040710.1 |
|
5 |
1822 |
|
GenBank mRNA |
AK124961.1 |
- |
5 |
1819 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR609457.1 |
- |
1 |
1703 |
|
Chr. 20 - 10023856 - 10026651 [-] |
View Full Product Details