Gene Transcripts

Gene Symbol
GLYATL1
Entrez Gene ID
92292
Gene Name
glycine-N-acyltransferase like 1
Gene Aliases
GATF-C, GNAT
Chromosome Location
Chr. 11: 58927629 - 58957074 on Build GRCh38
UniGene ID
Hs.38085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC013929.1 - 1 630

View Full Product Details