Gene Transcripts

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
Chromosome Location
Chr. 3: 129278806 - 129305293 on Build GRCh38
UniGene ID
Hs.458320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF035296.1 - 1 1476


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB559540.1 - 1 415 Chr. 3 - 129305751 - 129306183 [+]
GenBankmRNA DB568668.1 - 1 472 Chr. 3 - 129305694 - 129306183 [+]
GenBankmRNA BQ186731.1 - 1 512 Chr. 3 - 129305655 - 129306186 [+]
GenBankmRNA BM701712.1 - 1 497 Chr. 3 - 129305669 - 129306186 [+]
GenBankmRNA DB522972.1 - 1 452 Chr. 3 - 129305714 - 129306183 [+]
GenBankmRNA DB569505.1 - 1 452 Chr. 3 - 129305714 - 129306183 [+]
GenBankmRNA DB574025.1 - 1 442 Chr. 3 - 129305724 - 129306183 [+]
GenBankmRNA DB569457.1 - 1 449 Chr. 3 - 129305717 - 129306183 [+]
GenBankmRNA DB528801.1 - 1 446 Chr. 3 - 129305720 - 129306183 [+]
GenBankmRNA AA404698.1 - 1 334 Chr. 3 - 129305835 - 129306177 [+]

View Full Product Details