Gene Transcripts
- Gene Symbol
- C9orf3
- Entrez Gene ID
- 84909
- Gene Name
- chromosome 9 open reading frame 3
- Gene Aliases
- AOPEP, AP-O, APO, C90RF3, ONPEP
- Chromosome Location
- Chr. 9: 94726604 - 95148264 on Build GRCh38
- UniGene ID
- Hs.434253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042892.1 |
- |
1 |
1588 |
|
View Full Product Details