Gene Transcripts
- Gene Symbol
- LOC401433
- Entrez Gene ID
- 401433
- Gene Name
- uncharacterized LOC401433
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.647082
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127717.1 |
- |
1 |
2424 |
|
- Gene Symbol
- NUB1
- Entrez Gene ID
- 51667
- Gene Name
- negative regulator of ubiquitin like proteins 1
- Gene Aliases
- BS4, NUB1L, NYREN18
- Chromosome Location
- Chr. 7: 151341699 - 151378461 on Build GRCh38
- UniGene ID
- Hs.647082
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CN258712.1 |
- |
1 |
493 |
|
- Gene Symbol
- WDR86
- Entrez Gene ID
- 349136
- Gene Name
- WD repeat domain 86
- Gene Aliases
- Chromosome Location
- Chr. 7: 151377974 - 151410705 on Build GRCh38
- UniGene ID
- Hs.647083
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY304587.1 |
- |
2 |
479 |
|
Chr. 7 - 151377972 - 151404248 [-] |
View Full Product Details